Crystal violet advanced for Gram staining

Supplier: HARDY DIAGNOSTICS CA

C128A
CA89405-046EA 615.27 CAD
CA89405-046
Crystal violet advanced for Gram staining
Crystal Violet

Used as a primary stain in the advanced gram stain procedure


An aqueous solution of crystal violet.

Formula: C₂₅H₃₀ClN₃
MDL Number: MFCD00011750
CAS Number: 548-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR