Metro® Super Adjustable™ Wire Shelving, Chrome Plated, Metro™
Supplier: Metropolitan Wire
A1824NC A1830NC A1836NC A1842NC A1848NC A1854NC A1860NC A1872NC A2424NC A2430NC A2436NC A2442NC A2448NC A2454NC A2460NC A2472NC A1424NC A1430NC A1436NC A1442NC A1448NC A1460NC A1472NC A2124NC A2130NC A2136NC A2142NC A2148NC A2154NC A2160NC A2172NC A3036NC A3048NC A3060NC A3072NC A3636NC A3648NC A3660NC A3672NC A1424NC A1430NC A1436NC A1442NC A1448NC A1460NC A1472NC A2124NC A2130NC A2136NC A2142NC A2148NC A2154NC A2160NC A2172NC A3036NC A3048NC A3060NC A3072NC A3636NC A3648NC A3660NC A3672NC
CA22233-716EA 254.6 CAD
CA22233-716 CA22233-718 CA22233-720 CA22233-722 CA22233-724 CA22233-726 CA22233-728 CA22233-730 CA22233-732 CA22233-734 CA22233-736 CA22233-738 CA22233-740 CA22233-742 CA22233-744 CA22233-746 89369-754 89369-760 89369-766 89369-772 89369-778 89369-784 89369-790 89369-796 89369-802 89369-808 89369-814 89369-820 89369-826 89369-832 89369-838 89369-844 89369-848 89369-852 89369-856 89369-860 89369-864 89369-868 89369-872 CA89369-754 CA89369-760 CA89369-766 CA89369-772 CA89369-778 CA89369-784 CA89369-790 CA89369-796 CA89369-802 CA89369-808 CA89369-814 CA89369-820 CA89369-826 CA89369-832 CA89369-838 CA89369-844 CA89369-848 CA89369-852 CA89369-856 CA89369-860 CA89369-864 CA89369-868 CA89369-872
Metro® Super Adjustable™ Wire Shelving, Chrome Plated, Metro™
Furniture Shelves
Durable chrome-plated shelves are ideal for dry environments with minimal exposure to corrosives.
  • Design wire shelf system with appropriate accessories
An easily identifiable double groove is positioned every 20.3 cm (8") to further aid shelf alignment. Stationary posts are fitted with adjustable leveling bolts for use on uneven surfaces. Mobile posts require the purchase of stem casters. Stem casters (see 19766-236 series) and shelf dividers and ledges (see 22233-212 series) are available separately.
Notes: The actual length of the shelves is 3.2More Product Information