Contrex® AP Powder Detergent
Supplier: Decon Labs
5204 5225
CA89234-818CS 1852 CAD
CA89234-818 CA89234-820
Contrex® AP Powder Detergent
Cleaning Agents Detergents and Surface Cleansers
Contrex® AP is a powdered labware detergent.
  • Biodegradable and readily disposable
  • User friendly bottle allows for easy dispensing and eliminates caking
Contrex® AP removes virtually any contaminant including grease, oils, blood, tissue, particulates, deposits, chemicals, and solvents. It works by soaking, scrubbing, or in ultrasonic cleaning baths. The totally rinsable formulation eliminates detergent build-up that interferes with analysis, tissue culture, and other critical procedures. Its activity can be accelerated by heating, agitation, and
More Product Information