Eppendorf® Xplorer® Plus Electronic Multi-Channel Pipettes
Supplier: Eppendorf
4861000767 4861000775 4861000783 4861000791 4861000805 4861000813 4861000821 4861000830 4861000778 4861000779 4861000792 4861000793 4861000778 4861000779 4861000793
CA89234-008EA 2774.92 CAD
CA89234-008 CA89234-010 CA89234-012 CA89234-014 CA89234-016 CA89234-018 CA89234-020 CA89234-022 76311-130 76311-132 76311-134 76311-136 CA76311-130 CA76311-132 CA76311-136
Eppendorf® Xplorer® Plus Electronic Multi-Channel Pipettes
Pipettes Micropipettes
Eppendorf® Xplorer® plus multi-channel electronic pipettes offer the ideal solution for precise, efficient liquid handling in high-throughput applications, with ergonomic design, a user-friendly interface, and programmable modes for quick, reliable pipetting, enhancing reproducibility and minimizing errors.
  • Multifunctional rocker: 'Up is up and down is down™' principle
  • Clearly arranged color display; no sub-menus; help function
  • Innovative rocker: When the tip is ejected, the piston automatically returns to zero position
  • ON/OFF switch: Switches the pipetter
More Product Information