Microsep Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
MCP003C46 MCP030C46 MCPM02C68 MCP003C46 MCP010C41 MCP010C46 MCP030C41 MCP030C46 MCP100C41 MCP100C46 MCPM45C67 MCP003C41 MCPM02C67 MCPM45C67 MCPM45C68 MCP001C41 MCP001C46 MCPM02C68
89132-006PK 920.11 CAD
89132-006 89132-014 89132-022 CA89132-006 CA89132-008 CA89132-010 CA89132-012 CA89132-014 CA89132-016 CA89132-018 CA89132-024 CA89132-004 CA89132-020 89132-024 89132-026 CA89233-876 CA89233-878 CA89132-022
Microsep Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Filters Centrifugal Filters
Precise, quick recovery of microliter volumes of concentrate from starting volumes up to 5.0 ml.
  • Available with low protein-binding OmegaTM and Supor® (PES) membranes
  • Omega modified PES membranes come in a wide range of MWCOs, color-coded for easy identification
  • With Omega membranes you can Achieve 50X concentration and >90% recovery in just minutes
  • Supor (PES) membranes come in 0.45 and 0.2 µm pore sizes for microfiltration applications
  • Constructed of low-binding polypropylene
  • Ultrasonically welded seals prevent bypass or seal failure
For smallerMore Product Information