His Mag Sepharose® excel Magnetic Immobilized Metal Ion Affinity Chromatography (IMAC) Media, Cytiva
Supplier: Cytiva
17371220 17371221 17371222
CA89233-416EA 559.32 CAD
CA89233-416 CA89233-418 CA89233-420 89234-484 89234-486 89234-488 CA89234-484 CA89234-486 CA89234-488
His Mag Sepharose® excel Magnetic Immobilized Metal Ion Affinity Chromatography (IMAC) Media, Cytiva
Beads Magnetic Beads
His Mag Sepharose® excel is a magnetic immobilised metal ion affinity chromatography (IMAC) medium pre-charged with nickel ions that are strongly bound to a chelating ligand. The medium enables small scale capture, purification and reproducible screening of histidine-tagged proteins expressed and secreted into cell culture supernatants from eukaryotic cells such as insect cells or Chinese Hamster Ovary (CHO) cells.
  • Increased target protein yield
  • Minimised degradation through reduced and simplified sample handling
  • Simplify handling using visible and dense
More Product Information