Contrad® NF Non-foaming Liquid Detergent
Supplier: Decon Labs
6001 6002 6003 6055
CA89233-162CS 1121 CAD
CA89233-162 CA89233-164 CA89233-166 CA89233-168
Contrad® NF Non-foaming Liquid Detergent
Cleaning Agents Detergents and Surface Cleansers
Contrad® NF is a non-foaming concentrated liquid detergent designed for high-pressure cleaning applications in laboratory and industrial automatic washing machines, CIP systems, steam washers, etc.
  • Phosphate-free and chlorine-free
Contrad® NF is particularly recommended for critical cleaning of labware, where total rinsability and residue-free results are vital. It is also suitable for plant cleaning, such as biotechnology and pharmaceutical production equipment.