Combitips® advanced Positive Displacement Pipette Tips, Biopur® Individually Blister-Wrapped
Supplier: Eppendorf
0030089618 0030089626 0030089634 0030089642 0030089650 0030089669 0030089677 0030089685 0030089693
CA89232-966CS 333.54 CAD
CA89232-966 CA89232-968 CA89232-970 CA89232-972 CA89232-974 CA89232-976 CA89232-978 CA89232-980 CA89232-982
Combitips® advanced Positive Displacement Pipette Tips, Biopur® Individually Blister-Wrapped
Pipette Tips
Eppendorf Combitips® advanced, individually blister-wrapped in 'Biopur®' grade, offer unparalleled purity for critical lab tasks, ensuring contamination-free, precise pipetting with an ergonomic design. Ideal for highly sensitive molecular biology applications.
  • Combitips® advanced tips are the dedicated consumable for all Repeater®/Multipette® multi-dispenser pipettes from Eppendorf
  • Nine volume sizes (0.1 to 50 ml) offer a maximum range of dispensing volumes with increments as low as 100 nanoliters
  • Combitips® advanced in 'Biopur' quality are guaranteed to
More Product Information