PHCbi MPR Series Biomedical Refrigerators with Freezer Combo, PHC Corporation
Supplier: PHC Corporation
MPR-715F-PA MPR-715F-PA
89230-124EA 20054.47 CAD
89230-124 CA89230-124
PHCbi MPR Series Biomedical Refrigerators with Freezer Combo, PHC Corporation
Refrigerators
Ideal for biological, reagents, and vaccine storage, the MPR Series biomedical refrigerators with freezers combine high performance refrigeration and control and alarm/monitoring systems with energy-efficient, cost-effective cabinet design.
  • Uniformity with forced air circulation
  • Sample security with integrated alarm functions
  • Safe and secured storage behind a keyed locking door
  • Temperature controls and sensors
  • Separate refrigeration systems and compressors
  • Calibration adjustment through the control panel
Fan-forced air circulation in the refrigeratorMore Product Information