PURELAB® flex 3 Water Purification Systems, ELGA LabWater
Supplier: ELGA CA
SK-FLEX3-KIT SK-PF3XXXXM1-KIT SK-CSPFLEX-C SK-CSPFLEX-D SK-CSPFLEX-E SK-CSPFLEX-F SK-LPMAFLEX-34 SK-LPMAFLEX-34
89221-842EA 19094.66 CAD
89221-842 89221-844 CA89221-842 CA89221-844 76516-402 76516-404 76516-406 76516-408 76670-680 CA76670-680
PURELAB® flex 3 Water Purification Systems, ELGA LabWater
Water Purification Systems Pure and Ultra Pure Water Systems
The award winning PURELAB flex range is designed around your needs, delivering accuracy, flexibility and ease of use with an innovative and ergonomic design
  • The compact unit can be placed on the bench or can be wall mounted
  • Integrated 7 liter reservoir, ensuring that water is always available
  • The front doors ensure easy access to the consumables, making them quick and easy to replace
  • Data capture via USB for system performance validation and software updates
  • Fully recirculating
  • Ensuring the highest microbial purity and guaranteeing pure water at the point-
More Product Information