Versaflo™ S-Series Economy Hoods, 3M™
Supplier: 3M
7000127463 7000052786
CA89205-948CS 2335.13 CAD
CA89205-948 CA89205-950
Versaflo™ S-Series Economy Hoods, 3M™
Respirators Respiratory Systems
Helps provide head, face, neck and shoulder coverage.
  • Provides head, face, neck and shoulder coverage
  • For applications that require frequent replacement of entire headgear assembly
  • Fully assembled
  • OSHA APF 1000
Can be suitable for applications that require frequent replacement of the entire headgear assembly, such as pharmaceutical manufacturing, or in situations where there is less soiling of the outer fabric.
Fully assembled and ready to use out of the box. Color: white.
Certifications: OSHA APF 1000
Ordering information: Includes hood,More Product Information