Nunc® Cryobank and Bank-It™ Cryogenic Storage Vials, Thermo Scientific
Supplier: Thermo Fisher Scientific
374074 374088 374078 374080 374086 374258 374259 374260 374261 374081
CA82030-514CS 3007.09 CAD
CA82030-514 CA82030-518 CA82030-522 CA82030-526 CA82030-510 CA89202-186 CA89202-188 CA89202-190 CA89202-192 89238-102
Nunc® Cryobank and Bank-It™ Cryogenic Storage Vials, Thermo Scientific
Vials
Vials are designed for low-temperature storage of biological materials.
  • Features low protein adsorption, low DNA adsorption
  • Ideal for cells and proteins, storage of DNA
  • Sterility Assurance Level (SAL) of 10⁻⁶
  • Pyrogen-free according to the LAL-test
Vials are also available with unique 14×14 array 2D coding. The 2D code is laser-etched into the polypropylene bottom of the vial and cannot be lost during storage. The cryogenic storage box (82030-530) is constructed of white cardboard and holds 169 vials in a 13×13 format.
Vials snap into a 96-placeMore Product Information