Spectrum Two™ FT-IR Spectrometer and Analysis Systems, PerkinElmer
Supplier: PerkinElmer
L160000V L160000T L160000W L160000Q L160000R L160000S L160000F L160000U L160000A L1600114 L1600129 L1608009 L1600116 L1608010 L1600115 L1608012 L1608007 L1608006 L1608003 L1608004 L1608005 L1608022 L1608013 L1608001 L160000F L160000V L160000T L160000W L160000R L160000U L160000A L160000Q L160000S L1600129
CA89187-864EA 85013.45 CAD
CA89187-864 CA89187-866 CA89187-870 CA89187-878 CA89187-882 CA89187-884 CA89187-894 CA89187-900 CA89187-908 CA89187-910 CA89187-912 CA89187-914 CA89187-916 CA89187-918 CA89187-920 CA89187-922 CA89187-924 CA89187-930 CA89187-938 CA89187-940 CA89187-942 CA89187-948 CA89187-952 CA89187-966 89187-894 89187-864 89187-866 89187-870 89187-882 89187-900 89187-908 89187-878 89187-884 89187-912
Spectrum Two™ FT-IR Spectrometer and Analysis Systems, PerkinElmer
Spectrophotometers and Photometers IR and FTIR Spectrophotometers
A single software platform incorporates all of the functions required for infrared analyses, complete with instrument control, data manipulation and analysis, and flexible report utilities.
  • DynaScan interferometer
  • KBr optics
  • OpticsGuard™ technology
  • Atmospheric Vapor Compensation™ (AVC)
  • Compact design
  • Ready-made protocols
  • Materials library
  • Many resource packs for compliance with different industries
Spectrum Two FTIR Analysis Systems include the Spectrum Two instrument, Spectrum 10 or 10ES (Enhanced Security) software and the appropriate applicationMore Product Information