VWR® Ergonomic Smooth Floor Mats, Blue
Supplier: VWR International
89185-122 89185-124 89185-126 89185-128 89185-132 89185-136 89185-158 89185-160 89185-162 89185-164 89185-168 89185-170 89185-172 89185-196 89185-200 89185-202 89185-206 CA89185-122 CA89185-124 CA89185-126 CA89185-128 CA89185-130 CA89185-158 CA89185-160 76318-978 76318-980 76318-982 76318-984 76318-986 76318-988 76318-990
89185-122EA 562.51 CAD
89185-122 89185-124 89185-126 89185-128 89185-132 89185-136 89185-158 89185-160 89185-162 89185-164 89185-168 89185-170 89185-172 89185-196 89185-200 89185-202 89185-206 CA89185-122 CA89185-124 CA89185-126 CA89185-128 CA89185-130 CA89185-158 CA89185-160 76318-978 76318-980 76318-982 76318-984 76318-986 76318-988 76318-990
VWR® Ergonomic Smooth Floor Mats, Blue
Floor Mats Traction and Anti-Fatigue Floor Mats
The VWR® Smooth Mat is a true anti-fatigue mat. This smooth mat has the comfort of a polyurethane mat and the added benefit of easy clean-ability due to its closed-cell/non-porous top surface.
  • Stimulate the body to increase blood circulation and alterness
  • Molded beveled edges eliminate tripping hazards
  • Silcone free, latex free
  • Antistatic and meets ISO Class 5 (Class 100) compliance
  • Easy-to-clean, closed-cell/nonporous top surface
  • Highly resistant to chemicals and UV light
Safety features include molded beveled edges that will never curl up,More Product Information