VWR® Ergonomic Bubble Floor Mats, Blue
Supplier: VWR International
89184-910 89184-912 89184-914 89184-916 89184-918 89184-920 89184-922 89184-924 89184-946 89184-948 89184-950 89184-952 89184-956 89184-958 89184-960 89184-984 CA89184-910 CA89184-916 CA89184-918 CA89184-950 CA89184-984 76318-842 76318-844 76318-846 76318-848 76318-850 76318-852 76318-854 76318-856 76318-858 76318-860 76318-862 76318-864 76318-866 76318-868 76318-870 76318-872 76318-874 76318-876 76318-878 76318-880 76318-882 76318-884 76318-886 76318-888 76318-890 76318-892 76318-894 76318-896 76318-898 76318-900 76318-902 76318-904 76319-112 76319-114 76319-116 76319-118
89184-910EA 687.98 CAD
89184-910 89184-912 89184-914 89184-916 89184-918 89184-920 89184-922 89184-924 89184-946 89184-948 89184-950 89184-952 89184-956 89184-958 89184-960 89184-984 CA89184-910 CA89184-916 CA89184-918 CA89184-950 CA89184-984 76318-842 76318-844 76318-846 76318-848 76318-850 76318-852 76318-854 76318-856 76318-858 76318-860 76318-862 76318-864 76318-866 76318-868 76318-870 76318-872 76318-874 76318-876 76318-878 76318-880 76318-882 76318-884 76318-886 76318-888 76318-890 76318-892 76318-894 76318-896 76318-898 76318-900 76318-902 76318-904 76319-112 76319-114 76319-116 76319-118
VWR® Ergonomic Bubble Floor Mats, Blue
Floor Mats Traction and Anti-Fatigue Floor Mats
The VWR® Bubble Mat is a true ergonomic mat. The VWR Bubble mat has the comfort of a polyurethane mat and the added benefit of easy clean-ability due to its closed-cell/non-porous top surface.
  • Stimulate the body to increase blood circulation and alertness
  • Molded beveled edges eliminate tripping hazards
  • Silcone free, latex free
  • Anti static and meets ISO Class 5 (Class 100) compliance
  • Easy-to-clean, closed-cell/nonporous top surface
  • Highly resistant to chemicals and UV light
Safety features include molded beveled edges that will never curl up,More Product Information