ERGOMAT® Hygiene Rubber Mats, ERGOMAT
Supplier: Ergomat
EH0204-BLU EH0205-BLU EH0207-BLU EH0208-BLU EH0209-BLU EH0210-BLU EH0211-BLU EH0212-BLU EH0213-BLU EH0214-BLU EH0215-BLU EH0216-BLU EH0217-BLU EH0218-BLU EH0219-BLU EH0220-BLU EH0303-BLU EH0305-BLU EH0306-BLU EH0307-BLU EH0308-BLU EH0309-BLU EH0310-BLU EH0311-BLU EH0312-BLU EH0313-BLU EH0314-BLU EH0315-BLU EH0316-BLU EH0317-BLU EH0318-BLU EH0319-BLU EH0320-BLU EH0404-BLU EH0405-BLU EH0406-BLU EH0407-BLU EH0408-BLU EH0409-BLU EH0410-BLU EH0411-BLU EH0412-BLU EH0413-BLU EH0414-BLU EH0415-BLU EH0416-BLU EH0417-BLU EH0418-BLU EH0419-BLU EH0420-BLU EH0203-BLU EH0206-BLU EH0304-BLU EH0204-BLU EH0205-BLU EH0207-BLU EH0208-BLU EH0209-BLU EH0210-BLU EH0211-BLU EH0212-BLU EH0213-BLU EH0214-BLU EH0215-BLU EH0216-BLU EH0217-BLU EH0218-BLU EH0219-BLU EH0220-BLU EH0303-BLU EH0305-BLU EH0306-BLU EH0307-BLU EH0308-BLU EH0309-BLU EH0310-BLU EH0311-BLU EH0312-BLU EH0313-BLU EH0314-BLU EH0315-BLU EH0316-BLU EH0317-BLU EH0318-BLU EH0319-BLU EH0320-BLU EH0404-BLU EH0405-BLU EH0406-BLU EH0407-BLU EH0408-BLU EH0409-BLU EH0410-BLU EH0411-BLU EH0412-BLU EH0413-BLU EH0414-BLU EH0415-BLU EH0416-BLU EH0417-BLU EH0418-BLU EH0419-BLU EH0420-BLU
89169-608EA 611.11 CAD
89169-608 89169-610 89169-612 89169-614 89169-616 89169-618 89169-620 89169-622 89169-624 89169-626 89169-628 89169-630 89169-632 89169-634 89169-636 89169-638 89169-640 89169-642 89169-644 89169-646 89169-648 89169-650 89169-652 89169-654 89169-656 89169-658 89169-660 89169-662 89169-664 89169-666 89169-668 89169-670 89169-672 89169-674 89169-676 89169-678 89169-680 89169-682 89169-684 89169-686 89169-688 89169-690 89169-692 89169-694 89169-696 89169-698 89169-700 89169-702 89169-704 89169-706 CA22231-150 CA22231-152 CA22231-154 CA89169-608 CA89169-610 CA89169-612 CA89169-614 CA89169-616 CA89169-618 CA89169-620 CA89169-622 CA89169-624 CA89169-626 CA89169-628 CA89169-630 CA89169-632 CA89169-634 CA89169-636 CA89169-638 CA89169-640 CA89169-642 CA89169-644 CA89169-646 CA89169-648 CA89169-650 CA89169-652 CA89169-654 CA89169-656 CA89169-658 CA89169-660 CA89169-662 CA89169-664 CA89169-666 CA89169-668 CA89169-670 CA89169-672 CA89169-674 CA89169-676 CA89169-678 CA89169-680 CA89169-682 CA89169-684 CA89169-686 CA89169-688 CA89169-690 CA89169-692 CA89169-694 CA89169-696 CA89169-698 CA89169-700 CA89169-702 CA89169-704 CA89169-706
ERGOMAT® Hygiene Rubber Mats, ERGOMAT
Floor Mats Traction and Anti-Fatigue Floor Mats
The ERGOMAT® Hygiene is a cone profile ergonomic mat that is well suited for use in critical environments requiring daily caustic sanitation.
  • Hygiene mats have a broad range of pH resistance (2 to 13), and tolerate a variety of chemical exposures (except oils)
  • Mats can be used in hospitals and clinics, laboratories, food processing facilities, and commercial kitchens
  • Available in 4 colors (blue, gray, red, and green) to help eliminate the potential for cross contamination issues
  • Mats can be safely autoclaved (2’×3’ size only)
Mats are 11 mm (7/16")More Product Information