ERGOMAT® Hygiene Rubber Floor Mats, ERGOMAT
Supplier: Ergomat
EH0204-GRA EH0205-GRA EH0207-GRA EH0208-GRA EH0209-GRA EH0210-GRA EH0211-GRA EH0212-GRA EH0213-GRA EH0214-GRA EH0215-GRA EH0216-GRA EH0217-GRA EH0218-GRA EH0219-GRA EH0220-GRA EH0303-GRA EH0305-GRA EH0306-GRA EH0307-GRA EH0308-GRA EH0309-GRA EH0310-GRA EH0311-GRA EH0312-GRA EH0313-GRA EH0314-GRA EH0315-GRA EH0316-GRA EH0317-GRA EH0318-GRA EH0319-GRA EH0320-GRA EH0404-GRA EH0405-GRA EH0406-GRA EH0407-GRA EH0408-GRA EH0409-GRA EH0410-GRA EH0411-GRA EH0412-GRA EH0413-GRA EH0414-GRA EH0415-GRA EH0416-GRA EH0417-GRA EH0418-GRA EH0419-GRA EH0420-GRA EH0203-GRA EH0206-GRA EH0304-GRA EH0306-GRA EH0204-GRA EH0205-GRA EH0207-GRA EH0208-GRA EH0209-GRA EH0210-GRA EH0211-GRA EH0212-GRA EH0213-GRA EH0214-GRA EH0215-GRA EH0216-GRA EH0217-GRA EH0218-GRA EH0219-GRA EH0220-GRA EH0303-GRA EH0305-GRA EH0307-GRA EH0308-GRA EH0309-GRA EH0310-GRA EH0311-GRA EH0312-GRA EH0313-GRA EH0314-GRA EH0315-GRA EH0316-GRA EH0317-GRA EH0318-GRA EH0319-GRA EH0320-GRA EH0404-GRA EH0405-GRA EH0406-GRA EH0407-GRA EH0408-GRA EH0409-GRA EH0410-GRA EH0411-GRA EH0412-GRA EH0413-GRA EH0414-GRA EH0415-GRA EH0416-GRA EH0417-GRA EH0418-GRA EH0419-GRA EH0420-GRA
89169-308EA 616.97 CAD
89169-308 89169-310 89169-312 89169-314 89169-316 89169-318 89169-320 89169-322 89169-324 89169-326 89169-328 89169-330 89169-332 89169-334 89169-336 89169-338 89169-340 89169-342 89169-344 89169-346 89169-348 89169-350 89169-352 89169-354 89169-356 89169-358 89169-360 89169-362 89169-364 89169-366 89169-368 89169-370 89169-372 89169-374 89169-376 89169-378 89169-380 89169-382 89169-384 89169-386 89169-388 89169-390 89169-392 89169-394 89169-396 89169-398 89169-400 89169-402 89169-404 89169-406 CA22231-144 CA22231-146 CA22231-148 CA89169-344 CA89169-308 CA89169-310 CA89169-312 CA89169-314 CA89169-316 CA89169-318 CA89169-320 CA89169-322 CA89169-324 CA89169-326 CA89169-328 CA89169-330 CA89169-332 CA89169-334 CA89169-336 CA89169-338 CA89169-340 CA89169-342 CA89169-346 CA89169-348 CA89169-350 CA89169-352 CA89169-354 CA89169-356 CA89169-358 CA89169-360 CA89169-362 CA89169-364 CA89169-366 CA89169-368 CA89169-370 CA89169-372 CA89169-374 CA89169-376 CA89169-378 CA89169-380 CA89169-382 CA89169-384 CA89169-386 CA89169-388 CA89169-390 CA89169-392 CA89169-394 CA89169-396 CA89169-398 CA89169-400 CA89169-402 CA89169-404 CA89169-406
ERGOMAT® Hygiene Rubber Floor Mats, ERGOMAT
Floor Mats Traction and Anti-Fatigue Floor Mats
The ERGOMAT® Hygiene is a cone profile ergonomic mat well suited for use in critical environments requiring daily caustic sanitation
  • Use in hospitals and clinics, laboratories, food processing facilities, commercial kitchens
  • Available in 4 colors (blue, gray, red, and green) to help eliminate the potential for cross contamination issues
  • Mats can be safely autoclaved (2’ x 3’ size only)
  • Mats come with manufacturer’s three-year warranty
Mats are 11 mm (7/16") thick and 91.4 cm (36") wide, with beveled edges on all sides to prevent curling and eliminateMore Product Information