ERGOMAT® Softline® Rubber Mats, ERGOMAT
Supplier: Ergomat
SL0205-BK SL0207-BK SL0208-BK SL0209-BK SL0210-BK SL0211-BK SL0212-BK SL0213-BK SL0214-BK SL0215-BK SL0216-BK SL0217-BK SL0218-BK SL0219-BK SL0220-BK SL0305-BK SL0306-BK SL0307-BK SL0308-BK SL0309-BK SL0310-BK SL0311-BK SL0312-BK SL0313-BK SL0314-BK SL0315-BK SL0316-BK SL0317-BK SL0318-BK SL0319-BK SL0320-BK SL0404-BK SL0405-BK SL0406-BK SL0407-BK SL0408-BK SL0409-BK SL0410-BK SL0411-BK SL0412-BK SL0413-BK SL0414-BK SL0415-BK SL0416-BK SL0417-BK SL0418-BK SL0419-BK SL0420-BK SL0203 SL0204 SL0206 SL0303 SL0304 SL0207 SL0208 SL0209 SL0210 SL0211 SL0212 SL0213 SL0214 SL0215 SL0216 SL0217 SL0218 SL0219 SL0220 SL0306 SL0307 SL0308 SL0309 SL0310 SL0311 SL0312 SL0313 SL0314 SL0315 SL0316 SL0317 SL0318 SL0319 SL0320 SL0404 SL0405 SL0406 SL0407 SL0408 SL0409 SL0410 SL0411 SL0412 SL0413 SL0414 SL0415 SL0416 SL0417 SL0418 SL0419 SL0420
89169-112EA 787.31 CAD
89169-112 89169-114 89169-116 89169-118 89169-120 89169-122 89169-124 89169-126 89169-128 89169-130 89169-132 89169-134 89169-136 89169-138 89169-140 89169-142 89169-144 89169-146 89169-148 89169-150 89169-152 89169-154 89169-156 89169-158 89169-160 89169-162 89169-164 89169-166 89169-168 89169-170 89169-172 89169-174 89169-176 89169-178 89169-180 89169-182 89169-184 89169-186 89169-188 89169-190 89169-192 89169-194 89169-196 89169-198 89169-200 89169-202 89169-204 89169-206 CA22230-000 CA22230-002 CA22230-004 CA22231-046 CA22231-048 CA89169-114 CA89169-116 CA89169-118 CA89169-120 CA89169-122 CA89169-124 CA89169-126 CA89169-128 CA89169-130 CA89169-132 CA89169-134 CA89169-136 CA89169-138 CA89169-140 CA89169-144 CA89169-146 CA89169-148 CA89169-150 CA89169-152 CA89169-154 CA89169-156 CA89169-158 CA89169-160 CA89169-162 CA89169-164 CA89169-166 CA89169-168 CA89169-170 CA89169-172 CA89169-174 CA89169-176 CA89169-178 CA89169-180 CA89169-182 CA89169-184 CA89169-186 CA89169-188 CA89169-190 CA89169-192 CA89169-194 CA89169-196 CA89169-198 CA89169-200 CA89169-202 CA89169-204 CA89169-206
ERGOMAT® Softline® Rubber Mats, ERGOMAT
Floor Mats Traction and Anti-Fatigue Floor Mats
Pure nitrile rubber mats are ideal for areas exposed to significant moisture and slip hazards and are typically used in metal and welding operations, heavy manufacturing, and commercial kitchens.
  • Latex-free, nonporous, chemically resistant mats
  • Heat and abrasion resistant, and nonabsorbent
  • Withstand, animal grease, gasoline, oil, and organic solvents
These nonslip mats promote muscle stimulation, even while standing, which increases circulation and reduces fatigue and long-term injury. Mats are autoclavable at 121 °C (250 °F), or may be easily cleanedMore Product Information