ERGOMAT® Classic Antifatigue Floor Mats, ERGOMAT®
Supplier: Ergomat
ST0207 ST0209 ST0211 ST0213 ST0214 ST0215 ST0216 ST0217 ST0218 ST0219 ST0307 ST0309 ST0311 ST0313 ST0314 ST0315 ST0316 ST0317 ST0318 ST0319 ST0405 ST0407 ST0409 ST0411 ST0413 ST0414 ST0415 ST0416 ST0417 ST0418 ST0419 ST0203 ST0206 ST0304 ST0306 ST0404 ST0406 ST0204 ST0212 ST0308 ST0408 ST0410 ST0412 ST0420 ST0430 ST0460 ST0303 ST0205 ST0208 ST0305 ST0210 ST0330 ST0230 ST0220 ST0260 ST0310 ST0312 ST0320 ST0360 ST0207 ST0209 ST0211 ST0213 ST0214 ST0215 ST0216 ST0217 ST0218 ST0219 ST0307 ST0309 ST0311 ST0313 ST0314 ST0315 ST0316 ST0317 ST0318 ST0319 ST0405 ST0407 ST0409 ST0411 ST0413 ST0414 ST0415 ST0416 ST0417 ST0418 ST0419
89169-050EA 986.71 CAD
89169-050 89169-052 89169-054 89169-056 89169-058 89169-060 89169-062 89169-064 89169-066 89169-068 89169-070 89169-072 89169-074 89169-076 89169-078 89169-080 89169-082 89169-084 89169-086 89169-088 89169-090 89169-092 89169-094 89169-096 89169-098 89169-100 89169-102 89169-104 89169-106 89169-108 89169-110 CA22231-010 CA22231-012 CA22231-016 CA22231-018 CA22231-020 CA22231-022 CA22231-024 CA22231-028 CA22231-032 CA22231-120 CA22231-122 CA22231-124 CA22231-126 CA22231-128 CA22231-130 CA22231-014 CA22231-026 CA22231-027 CA22231-030 CA22231-034 CA22231-036 CA22231-038 CA22231-106 CA22231-108 CA22231-110 CA22231-112 CA22231-114 CA22231-116 CA89169-050 CA89169-052 CA89169-054 CA89169-056 CA89169-058 CA89169-060 CA89169-062 CA89169-064 CA89169-066 CA89169-068 CA89169-070 CA89169-072 CA89169-074 CA89169-076 CA89169-078 CA89169-080 CA89169-082 CA89169-084 CA89169-086 CA89169-088 CA89169-090 CA89169-092 CA89169-094 CA89169-096 CA89169-098 CA89169-100 CA89169-102 CA89169-104 CA89169-106 CA89169-108 CA89169-110
ERGOMAT® Classic Antifatigue Floor Mats, ERGOMAT®
Floor Mats Traction and Anti-Fatigue Floor Mats
Durable, easy to clean, and shock absorbent, these polyurethane floor mats offer outstanding ergonomic benefits
  • Durable, easy to clean, shock absorbent, static dissipative
  • For use in ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanrooms
  • Ideal for dry, nonabrasive areas
  • 1.6 cm (5/8") thick with four beveled edges
Designed to reduce fatigue and combat the cumulative stress and anatomic discomforts associated with foot, leg, and back fatigue from extensive standing. Mats are static dissipative, suitable for use in ISO Class 5 (FED-STD-209E Class 100/M3.5)More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable