Well-Coated™ Amine Binding for Protein/Peptide Binding, G-Biosciences
Supplier: G-Biosciences
786-753 786-756 786-757
CA89167-978EA 656.93 CAD
CA89167-978 CA89167-984 CA89167-986
Well-Coated™ Amine Binding for Protein/Peptide Binding, G-Biosciences
Microplates HTS Microplates
G-Biosciences' Well-Coated™ Amine Binding plates are designed to specifically bind primary amines of peptides, proteins and other molecules
G-Biosciences' Well-Coated™ Amine Binding plates are maleic anhydride activated plates that react with primary amines to form amide bonds that are stable at pH≥7. Acidic conditions will hydrolyze the bonds releasing the peptide/ligand, therefore binding of peptide/ligand to plates should be performed at pH8-9 and the binding assays or ELISA should be performed at pH≥7.
The wells are coated to a 200µL depth and are supplied pre-blocked in ourMore Product Information