Well-Coated™ Protein L Plates for Antibody Binding, G-Biosciences
Supplier: G-Biosciences
786-737 786-776 786-777
CA89167-946EA 1112.51 CAD
CA89167-946 CA89168-024 CA89168-026
Well-Coated™ Protein L Plates for Antibody Binding, G-Biosciences
Microplates HTS Microplates
G-Biosciences' Well-Coated™ Protein L plates are designed to bind the kappa light chains of immunoglobulins without interfering with the antigen binding site.
G-Biosciences' Well-Coated™ Protein L plates offer greater range of immunoglobulin classes and subclasses compared to Protein A, Protein G and Protein A/G. Protein L will bind to all classes of IgG, including IgG, IgM, IgA, IgE and IgD, and binds to single chain variable fragments (scFv and Fab fragments).
G-Biosciences' Well-Coated™ Protein L plates are for single antibody assays and are not suitable for multiple assays (More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable