Well-Coated™ Protein G Plates for Antibody Binding, G-Biosciences
Supplier: G-Biosciences
786-733 786-774 786-775
CA89167-938EA 1052.36 CAD
CA89167-938 CA89168-020 CA89168-022
Well-Coated™ Protein G Plates for Antibody Binding, G-Biosciences
Microplates HTS Microplates
G-Biosciences' Well-Coated™ Protein G plates are designed to bind the constant (Fc) region of immunoglobulins, ensuring that the antigen binding domain of the antibody is orientated away from the plate, offering maximum exposure of the binding site.
The immunoglobulin orientation on the Well-Coated™ Protein G plates improves the antibody capacity compared to plates that are coated directly with antibodies.
G-Biosciences' Well-Coated™ Protein plates are for single antibody assays and are not suitable for multiple assays (sandwich ELISAs) as the first antibody will not block all IgGMore Product Information