Tube-O-CONCENTRATOR™ Protein Concentrator, G-Biosciences
Supplier: G-Biosciences
786-625 786-626
CA89167-814EA 674.6 CAD
CA89167-814 CA89167-816
Tube-O-CONCENTRATOR™ Protein Concentrator, G-Biosciences
Tubes Dialysis Tubes
Tube-O-CONCENTRATOR™ is a versatile concentration device that utilizes a novel, water absorbing, liquid polymer and our patented Tube-O-DIALYZER™ for the rapid concentration of dilute protein solutions with zero protein loss
The Tube-O-CONCENTRATOR™ solution is a liquid polymer that rapidly absorbs water through the dialysis membrane in the Tube-O-DIALYZER™ cap, which retains all molecules with a molecular weight >1kDa.
Tube-O-CONCENTRATOR™ is available in two sizes for concentrating sample volumes of up to 250 µl (Micro) or 2.5 ml (Medi).
The unique tube design of Tube-O-More Product Information