DuPont™ Tychem® 2000 Coveralls with Standard Hood and Attached Socks, Taped Seams
Supplier: DuPont
QC122TYLMD000400 QC122TYLLG000400 QC122TYLXL000400 QC122TYL2X000400 QC122TYL3X000400 QC122TYL4X000400 QC122TYL5X000400 QC122TYL6X000400
CA89138-050CS 955.75 CAD
CA89138-050 CA89138-052 CA89138-054 CA89138-056 CA89138-058 CA89138-060 CA89138-062 10016-394
DuPont™ Tychem® 2000 Coveralls with Standard Hood and Attached Socks, Taped Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Standard fit hood
  • Elastic wrists
  • Attached socks
  • Storm flap with adhesive closure
  • Taped seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Taped seamsMore Product Information