CoreDish™ Multiple Biopsy Containers, Simport Scientific
Supplier: Simport Scientific
M970-D5B-2 M970-D8P M970-D8UGI M970-D12LGI M970-D12P M970-D5 M970-D8 M970-D12 M971-D8P M971-D12P M970-D5B-1 M971-D5B-2 M971-D8UGI M971-D12LGI M971-D5 M971-D8 M971-D12 M970-D5B-1
CA89137-774CS 112.36 CAD
CA89137-774 CA89137-776 CA89137-778 CA89137-780 CA89137-782 CA89137-784 CA89137-786 CA89137-788 CA89176-826 CA89176-832 CA97060-900 CA11028-504 CA11028-506 CA11028-508 CA11028-510 CA11028-512 CA11028-514 76299-264
CoreDish™ Multiple Biopsy Containers, Simport Scientific
Embedding Cassettes Biopsy Processing Cassettes
These polystyrene containers are half-filled with 10% neutral buffered formalin
The CoreDish® is available half prefilled with 10% neutral buffered formalin or empty for more versatility.
They feature leakproof closures with an O-ring, and clearly marked compartments. A label allows essential information to be written on the base. Different configurations are available for general purposes, breast, prostate, upper GI, and lower GI biopsies
Certifications: CE Marked