AcroPrep™ Filter Plates For Ultrafiltration, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
5080 5081 5076 5077 5078 5079 8163 8164 8166 8033 8034 8035 8036 8165
CA10278-002PK 1242.44 CAD
CA10278-002 CA10278-004 CA28148-670 CA28148-672 CA28148-674 CA28148-676 CA89135-696 CA89135-698 CA89135-702 CA97052-106 CA97052-108 CA97052-110 CA97052-112 CA89135-700
AcroPrep™ Filter Plates For Ultrafiltration, Cytiva (Formerly Pall Lab)
Microplates Filter Plates
AcroPrep and AcroPrep Advance filter plates combine high performance Omega membranes with unique well designs to provide rapid, efficient separation of biomolecules. These plates are suitable for applications such as size exclusion, PCR clean-up, nucleic acid purification, and protein separation.
  • High performance Omega membrane provides high recovery and typically results in ≥ 90% recovery of target biomolecules
  • AcroPrep Plates eliminate crosstalk between wells, membranes are individually sealed in each well with a proprietary sealing process and each well
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Well bottom area 0.05 cm² 0.25 cm²