Corning® Human Recombinant Albumin (from Serum)
Supplier: Corning
62-450-RF
CA89133-078EA 182.09 CAD
CA89133-078
Corning® Human Recombinant Albumin (from Serum)
Proteins and Peptides
Corning® cellgro® Albumin is a chemically-defined recombinant human serum albumin (rhHSA). Having an essential and non-essential fatty acid profile, this is a powerful media supplement that outperforms plasma-derived human serum albumin (pHSA), bovine serum albumin (BSA) and FBS. Some prominent applications for cellgro® rhAlbumin are as a supplement for CHO cell culture and for the culture of various other cell lines. This product is plant-derived and completely animal-origin free.