TWIRL'EM ECOLO Biodegradable Sampling Bags, Labplas
Supplier: Labplas
EPL-3070E EPL-4590E EPL-5590E EPL45590E EPL-5515E EPL-7012E EPL47012E EPL-7015E EPR-3050E EPR-3070E EPR-4590E EPR-5590E EPR-7012E EPR47012E EFL-4590E EFL-5590E EFL-5512E EFL-7012E EFL47012E EFL-1012E EFL-1015E EFR-3070E EFR-4590E EFR-5590E EFR-7012E EFR47012E EFR-1012E EFR-1015E
CA89131-780CS 5093.76 CAD
CA89131-780 CA89131-782 CA89131-784 CA89131-786 CA89131-788 CA89131-790 CA89131-792 CA89131-794 CA89131-796 CA89131-798 CA89131-800 CA89131-802 CA89131-804 CA89131-806 CA89131-808 CA89131-810 CA89131-812 CA89131-814 CA89131-816 CA89131-818 CA89131-820 CA89131-822 CA89131-824 CA89131-826 CA89131-828 CA89131-830 CA89131-832 CA89131-834 CA89131-836
TWIRL'EM ECOLO Biodegradable Sampling Bags, Labplas
Bags Sample Bags
These biodegradable sample bags are ideal for transportation and storage of solids, semisolids, and liquids for environmental and carcass sampling, biomedical and pharmaceutical research, quality assurance procedures, food industry applications, and clinical and veterinary medicine.
  • Made of pliant, highly resistant virgin polyethylene sterile tubing with no side seals
  • Feature a wire closure for an airtight seal
  • Wide-sealed bottoms for added leakproof protection
They are easy to use, tear off perforated top, pull tabs to open bag, fill, grab ends ofMore Product Information