JACKSON SAFETY® V30 Nemesis™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Ansell Healthcare
25685 25685
89130-928EA 19.19 CAD
89130-928 CA89130-928
JACKSON SAFETY® V30 Nemesis™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Safety Glasses
Sporty, flexible and lightweight design, the JACKSON SAFETY® V30 Nemesis safety eyewear have soft-touch temples for added comfort.
  • Base curve 8
  • Single-lens wraparound protection
Enhanced nosepiece design with integrated ridges to channel away sweat. Polycarbonate material that provides 99.9% UVA/UVB/UVC protection. Hardcoated lenses for scratch resistance and longer life.
Certifications: ANSI Z87.1-2010 standard.
Ordering information: Every pair includes a neck cord.
Order Now
SPECIFICATIONS
Environmentally Preferable