DuPont™ Tyvek® IsoClean® Coveralls with Dolman Sleeves and Snaps
Supplier: DuPont
IC254BWH2X0025CS IC254BWH3X0025CS IC254BWH4X0025CS IC254BWHLG0025CS IC254BWHMD0025CS IC254BWHSM0025CS IC254BWHXL0025CS
CA89129-662CS 4251.69 CAD
CA89129-662 CA89129-664 CA89129-666 CA89129-668 CA89129-670 CA89129-672 CA89129-674
DuPont™ Tyvek® IsoClean® Coveralls with Dolman Sleeves and Snaps
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Bound seams
  • Bound neck
  • Dolman sleeve design
  • Elastic wrists and ankles
  • Zipper closure
  • Snaps for aseptic donning
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration. Coveralls feature elastic openings at wrists and ankles for a tighter fit. Bound neck allowsMore Product Information