LockMailer™ Microscope Slide Jar, Simport Scientific
Supplier: Simport Scientific
M950-4MA M950-4MA
89129-506CS 512.32 CAD
89129-506 CA89129-506
LockMailer™ Microscope Slide Jar, Simport Scientific
Slide Mailers
Tamper-evident, multi-purpose container for mailing, staining, or storing microscope slides
  • Holds four standard microscope slides
  • Slotted container to keep slides separated
  • Optional unique tamper-evident cap
  • Different colors are available
Made of strong, clear polypropylene. It holds four standard slides vertically, and incorporates a unique tamper-evident, leakproof screw cap. Slots keep the slides from sticking together
Constructed of extra-strong and clear polypropylene. It will hold up to four standard 3×1" or 75×25 mm slides vertically. InsideMore Product Information