Finnpipette® F2 Single-Channel Fixed Volume Pipettors, Thermo Scientific
Supplier: Thermo Fisher Scientific
4652000 4652010 4652020 4652030 4652040 4652050 4652070 4652090 4652100 4652110 4652120 4652130 4652140 4652080 4652000 4652010 4652020 4652030 4652040 4652070 4652090 4652100 4652110 4652120 4652130 4652140
89080-366EA 727.41 CAD
89080-366 89080-368 89080-370 89080-372 89080-374 89080-376 89080-380 89080-384 89080-386 89080-388 89080-390 89126-254 89126-256 89080-382 CA89080-366 CA89080-368 CA89080-370 CA89080-372 CA89080-374 CA89080-380 CA89080-384 CA89080-386 CA89080-388 CA89080-390 CA89126-254 CA89126-256
Finnpipette® F2 Single-Channel Fixed Volume Pipettors, Thermo Scientific
Pipettes Micropipettes
Finnpipette® F2 fixed volume pipettors offer outstanding accuracy and precision in laboratory based liquid dispensing procedures.
  • Each lightweight device features an enlarged digital display
  • Offers remarkably smooth pipetting action
  • Pipettors are fully autoclavable and easy to service
Pipettors are equipped with an advanced volume gearing mechanism (AVG) designed to improve accuracy and precision.
These pipettors combine the established Finnpipette® features with state-of-the-art innovations to create a reliable and ergonomic high-performanceMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable