Eppendorf® Research® Plus Series Multichannel Pipettors
Supplier: Eppendorf
3125000010 3125000036 3125000052 3125000028 3125000060 3125000044 2231300043 2231300045 2231300047 2231300044 2231300048 3125000079 3125000087 3125000095 3125000109 3125000079 3125000087 3125000095 3125000109
CA89125-336EA 1872.69 CAD
CA89125-336 CA89125-338 CA89125-340 CA89125-342 CA89125-346 CA89125-344 CA490013-002 CA490013-004 CA490013-006 CA490013-008 CA490013-012 76311-122 76311-124 76311-126 76311-128 CA76311-122 CA76311-124 CA76311-126 CA76311-128
Eppendorf® Research® Plus Series Multichannel Pipettors
Pipettes Micropipettes
The Research® Plus series of adjustable volume multichannel pipettors are designed to accommodate the rapidly developing needs of laboratory researchers.
  • Color-coding on the operating button simplifies tip identification and selection
  • All replacement manifolds are sold separately
  • Fully autoclavable devices can be easily disassembled for cleaning and decontamination
  • Feature a specially designed, spring-loaded tip cone to help reduce the force required for tip attachment and ejection
The rounded and elongated grip fits comfortably in the hand, enhancingMore Product Information