ColorTouch® Pink Powder-Free Latex Examination Gloves, Microflex®, Ansell
Supplier: Microflex
CTP-233-M
CA89107-526BX 32.32 CAD
CA89107-526 CA89107-528 CA89107-530 CA89107-532
ColorTouch® Pink Powder-Free Latex Examination Gloves, Microflex®, Ansell
Gloves
Gloves deliver medical-grade barrier protection with consistent comfort and quality.
  • Ambidextrous, beaded cuff
  • Fully textured for a secure grip
  • For single use only
  • Thickness: 6.7 mil (finger)
High-quality, polymer-coated latex is easy to don and reduces hand fatigue.
Note: Donation of $1 for each case purchased is made to The Breast Cancer Research Foundation®.