Nunc® Cryobank Freezer Racks, Thermo Scientific
Supplier: Thermo Fisher Scientific
367014
CA89094-374EA 1163.16 CAD
CA89094-374
Nunc® Cryobank Freezer Racks, Thermo Scientific
Racks Cryogenic Box Racks
This 74cm rack holds nineteen 0.5mL or thirteen 1.0mL Cryobank racks.