Deepwell Plates 384-Well, 200 µl
Supplier: Eppendorf
951031003 951031402 951031101 951031500 0030113551
CA89085-658CS 780.02 CAD
CA89085-658 CA89085-660 CA89085-662 CA89085-664 CA77395-526
Deepwell Plates 384-Well, 200 µl
Microplates Deep Well Microplates
Eppendorf Deepwell plates 384-well, 200 µl offer efficient sample handling with features like OptiTrack® matrix for rapid identification and g-Safe® technology for stability during centrifugation. SafeCode variants provide secure three-level coding, including error correction for reliable identification, ensuring flexibility and accuracy in experimental workflows.
  • OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax® well design: Optimized well geometry for maximum sample
More Product Information