Proline® Plus Single Channel Mechanical Pipettors, Sartorius
Supplier: Sartorius
728020 728030 728050 728060 728070 728080 728515 728520 728530 728535 728545 728565 728575 728040 728010 728090 728550 728560 728567 728570 728580 728590 728010 728020 728030 728050 728060 728070 728080 728090 728515 728520 728530 728535 728545 728550 728560 728565 728567 728570 728575 728580 728590 728040 LH728670 LH728671 LH728672 LH728673 LH728674 LH728675 LH728676 LH728677
89082-276EA 839.98 CAD
89082-276 89082-278 89082-280 89082-282 89082-284 89082-286 89082-302 89082-304 89082-306 89082-308 89082-310 89082-316 89082-322 89177-094 89082-274 89082-288 89082-312 89082-314 89082-318 89082-320 89082-324 89082-326 CA89082-274 CA89082-276 CA89082-278 CA89082-280 CA89082-282 CA89082-284 CA89082-286 CA89082-288 CA89082-302 CA89082-304 CA89082-306 CA89082-308 CA89082-310 CA89082-312 CA89082-314 CA89082-316 CA89082-318 CA89082-320 CA89082-322 CA89082-324 CA89082-326 CA89177-094 75845-038 75845-040 75845-042 75845-044 75845-046 75845-048 75845-050 75845-052
Proline® Plus Single Channel Mechanical Pipettors, Sartorius
Pipettes Micropipettes
Proline® Plus mechanical pipettors are designed to offer comfort and quality for everyday manual pipetting.
  • Ergonomic design with light pipetting forces, comfortable handle, and finger support for reduced risk of strain injuries
  • Minimized risk of contamination with Safe-Cone Filters and full autoclavability
  • Highly durable with strengthened structure for heavier use
Pipettors offer good UV resistance and excellent chemical resistance. The devices are suitable for both left- and right-handed users. Thermal insulation of internal components ensures thatMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable