Zinc sulfate heptahydrate ≥99%, granular USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA
Danger

ZI120-12KG ZI120-50KGBL
CA700001-536EA 4340.01 CAD
CA700001-536 CA89079-058
Zinc sulfate heptahydrate ≥99%, granular USP
Zinc sulphate heptahydrate

BSE/TSE statement (available upon request) Allergen statement (available upon request)


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: ZnSO₄·7H₂O
MW: 287.54 g/mol
Melting Pt: 100
Density: 1.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9 III
Merck Index: 12,10293

Order Now

Specification Test Results

Assay 99% Min.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR