Zinc sulfate monohydrate ≥98% FCC suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP CA
Danger

Z1100-50KGBL
CA89079-056EA 9532 CAD
CA89079-056
Zinc sulfate monohydrate ≥98% FCC suitable for use as excipient
Zinc sulphate monohydrate

Zinc Sulfate, Monohydrate, FCC is used as a nutritional supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: ZnSO₄·1H₂O
MW: 179.45 g/mol
Density: 3.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149893
CAS Number: 7446-19-7
EINECS: 231-793-3
UN: 3077
ADR: 9 III
Merck Index: 13,10213

Order Now

Specification Test Results

Assay 98.0 - 100.5 %
ACIDITY TO PASS TEST
ALKALIES AND ALKALINE EARTHS max. 0.5 %
CADMIUM (Cd) max. 5 mg/kg
LEAD (Pb) max. 4 mg/kg
MERCURY max. 5 mg/kg
SELENIUM (Se) max. 0.003 %
IDENTIFICATION TO PASS TEST


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR