Mouse Igloo™ Enrichment Housings
Supplier: Bio-Serv
K3327 K3328 K3250 CA89067-856 K3327 K3328 K3250 K3570
CA89067-850EA 32.31 CAD
CA89067-850 CA89067-852 CA89067-854 CA89067-856 CA89067-858 89067-850 89067-852 89067-854 CA89412-072
Mouse Igloo™ Enrichment Housings
Animal Supplies Animal Environmental Enrichment
Transparent polycarbonate igloos are ideal for cage checks and breeding programs
Fast-Tracs are unique activity wheels for use with the mouse Igloo that support natural and instinctive running behavior. Igloo floor snaps onto the mouse Igloo and allows mother and pups to be transferred together, improving pup survival rate.
Allows mice to withdraw from direct light and seek refuge from dominant cage mates. Certified (contaminant-screened) and autoclavable.
Replacement pins: Acetal copolymer plastic pin connects fast-trac to Igloo.
Order Now
SPECIFICATIONS
Environmentally Preferable