Chain for Enrichment Devices, Bio-Serv
Supplier: Bio-Serv
9132
CA89067-734EA 16.72 CAD
CA89067-734
Chain for Enrichment Devices, Bio-Serv
Animal Supplies Animal Environmental Enrichment
All stainless steel welded-link chain
Hang on pens to encourage play behavior. Use with Quick Link or Twist Clip to make links. Custom lengths available. Autoclavable. Cagewasher safe.