Dustless Precision Pellets® Rodent Purified Diet, Bio-Serv
Supplier: Bio-Serv
F05684 F0071 F0021 F0078 F05301 F0299 F05684 F0021 F0078 F0299 F0075
CA89067-560EA 658.97 CAD
CA89067-560 CA89067-562 CA89067-564 CA89067-566 CA89067-574 CA89067-576 CA89067-578 89067-560 89067-564 89067-574 89067-578 CA89408-190
Dustless Precision Pellets® Rodent Purified Diet, Bio-Serv
Animal Supplies Animal Feed
Precise size and weight pellets generally used in nutritional and behavioral studies.
  • Dustless and fracture free
  • Superior performance in automatic feeders
  • Nutritionally complete purified formula
  • Wide range of weights: 14 mg to 1 g
  • Available in grain-based formula
  • Every batch tested for hardness, weight, and height
  • Custom flavors and colors available upon request
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage
Ingredients: Sucrose, Dextrose, Casein, Corn Oil, Mineral Mix, Cellulose, Corn Syrup, Calcium Silicate, Vitamin Mix, MagnesiumMore Product Information