Turf Foraging Crumbles™ Treats, Bio-Serv
Supplier: Bio-Serv
F05433 F05437
CA89067-396EA 287.75 CAD
CA89067-396 CA89067-398 CA89067-408 CA89067-410
Turf Foraging Crumbles™ Treats, Bio-Serv
Animal Supplies Animal Feed
Small, fruit-flavored bits, available in a variety of flavors, are nutritionally complete foraging treats
  • Specially sized for use with Bio-Serv's Turf Foraging Board
  • Scatter into bedding to encourage rooting behavior in swine
  • Saves labor and less expensive than perishable fruit
  • Uncolored Banana and Pina Colada Flavors are Certified (Contaminant Screened) and suitable for use in GLP and TOX studies
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage
Ingredients: Dextrose, Casein, Cellulose, Corn Starch, Sucrose, Mineral Mix, Canola Oil,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable