Supreme Mini-Treats™, Certified, Bio-Serv
Supplier: Bio-Serv
F05711-1 F05472-1 F05474-1 F05475-1 F05478-1 F05711 F05472 F05474 F05475 F05478 F05472-1 F05474-1 F05475-1 F05478-1 S05478-1 S05472-1 S05474-1 S05475-1 S05711-1
CA89067-266PK 221.06 CAD
CA89067-266 CA89067-268 CA89067-270 CA89067-272 CA89067-274 CA89067-276 CA89067-278 CA89067-280 CA89067-282 CA89067-284 89067-268 89067-270 89067-272 89067-274 CA89408-288 CA89408-290 CA89408-292 CA89408-294 CA89408-296
Supreme Mini-Treats™, Certified, Bio-Serv
Animal Supplies Animal Feed
Nutritionally complete 1 g treats that compliment enrichment programs. Excellent for training and conditioning or as a reward following a less desirable procedure.
  • Appetizing aroma helps to deter rodent maternal cannibalism
  • chocolate and very berry flavor recommended.
  • Excellent source of vitamin C
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage
Ingredients: Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corn Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Natural and Artificial Flavors, Vitamin Mix, Calcium Silicate, MagnesiumMore Product Information