tri-Sodium citrate dihydrate 99,0, granulated USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

SO165-12KG SO165-50KGBL
CA89050-470EA 3636.01 CAD
CA89050-470 75812-436
tri-Sodium citrate dihydrate 99,0, granulated USP
tri-Sodium citrate dihydrate

Sodium Citrate, Dihydrate, Granular, USP can be used as an alkalizing agent, buffering agent, emulsifier, or sequestering agent. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₆H₅Na₃O₇) 99.0 - 100.5 %
Water 10.0 - 13.0 %
Alkalinity To Pass Test
Tartrate To Pass Test
Heavy Metals max. 0.001 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR