KLEENGUARD® A70 Chemical Spray Protection Apron, Kimberly-Clark
Supplier: Microflex
97790
CA89030-872CS 359.61 CAD
CA89030-872
KLEENGUARD® A70 Chemical Spray Protection Apron, Kimberly-Clark
Disposable Garments
The KLEENGUARD® A70 line of apparel is designed to provide protection against chemical spray in hazardous waste remediation, environmental cleanup, petrochemical/oil refining, acid/caustic handling, biohazard cleanup, and pharmaceutical manufacturing applications
  • Seamless design
  • Soft and durable fabric
  • Wick-away lining for increased comfort
  • Neck loop and waist level ties provide secure fitting
NFPA 99 compliant for antistatic material.