Nitri-Knit™ Supported Nitrile Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
NK803/8-H5 NK803/11 NK803/7 NK803/9 NK803/10 NK803ES/7 NK803ES/8 NK803ES/9 NK803ES/11 NK803IN/7 NK803IN/8 NK803IN/9 NK803IN/10 NK803IN/11
CA11001-469PK 316.54 CAD
CA11001-469 CA57100-102 CA11001-527 CA57100-100 CA57100-104 CA57100-106 CA89092-984 CA89092-986 CA89092-988 CA89092-990 CA89022-154 CA89022-156 CA89022-158 CA89022-160 CA89022-162
Nitri-Knit™ Supported Nitrile Gloves, Honeywell Safety
Gloves
For use in a wide range of applications, including: automotive assembly and painting; chemical handling and processing; degreasing; dip tank cleanup; fishing industry processing; food processing; printing cleanup; and general refinery, solvent, and maintenance work.
  • High nitrile content offers superior resistance to cuts, snags, and punctures
  • Interlock knit liner adds strength and comfort
  • Treated with permanent antibacterial and anti-mildew agent
  • Color-coded stitching makes size identification fast and easy
  • Compliant with USDA and FDA CFR 21 food
More Product Information