A800 Series Protective Eyewear, Honeywell Safety
Supplier: Honeywell Safety Products
A800 A801 A805
CA89020-076CS 1684.22 CAD
CA89020-076 CA89020-078 CA89020-080 CA89020-082
A800 Series Protective Eyewear, Honeywell Safety
Safety Glasses
Stylish, sporty, and economic A800 series eyewear provides fashionable protection for the everyday wearer.
  • Sport-style temples feature soft, padded inserts with extra flex built in for more comfort during extended use
  • Soft nosepiece conforms to various nose bridge shapes for a nonslip fit
  • 9-base polycarbonate hardcoated lens provides a 180° field of distortion-free vision and provides 99.9% UV protection
The A800 series is available with a variety of lens types for both indoor and outdoor work applications. They are also available with either an anti-More Product Information