SAF-T-FIT™ Plus N1125 Disposable Respirators, Honeywell Safety
Supplier: Honeywell Safety Products
14110393 14110394 14110395 14110396 14110398 14110399 14110400 14110401
CA89013-600PK 89.1 CAD
CA89013-600 CA89013-602 CA89013-604 CA89013-606 CA89013-608 CA89013-610 CA89013-612 CA89013-614 CA89013-616 89013-612
SAF-T-FIT™ Plus N1125 Disposable Respirators, Honeywell Safety
Respirators Disposable Respirators Particulate Respirators
Ideal for welding, soldering, brazing, torch cutting, metal pouring, grinding, sanding, sweeping, or other hot and dusty operations. Respirators feature an adjustable boomerang nose seal for a secure fit and added comfort. Nose seal is color-coded for easy size recognition. Respirators are equipped with an exhalation valve for ease of breathing and exceptional comfort during extended wear.
  • Offers latex-free N95 protection from particulate aerosols free of oil
  • Exhalation valve improves comfort during extended wear
  • Adjustable boomerang nose seal fits
More Product Information