Nonconductive Safety Padlocks, with 1.5" Shackle, Brady Worldwide®
Supplier: Brady Worldwide
99552 99556 99564 99570 99576 99580
CA89008-058EA 22.33 CAD
CA89008-058 CA89008-060 CA89008-062 CA89008-064 CA89008-066 CA89008-068
Nonconductive Safety Padlocks, with 1.5" Shackle, Brady Worldwide®
Security Locks
Nonconductive Safety Padlocks are the ultimate locks for all safety and security needs
The unique cylinder is insulated to protect workers from shocks when key is inserted. The cylinder is six-pin precision-machined for more unique key cuts and improved tamper resistance. Reserved, paracentric keyway provides optimal security and the special design prohibits the key from being released until the padlock shackle is closed. Locks are compact and lightweight, and they resist impact, temperature, chemicals, and corrosion. Their nonconductive, nonsparking body features a ribbed design forMore Product Information